Ethyl decanoate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Ethyl caprate, Capric acid ethyl ester, Decanoic acid ethyl ester, Decylic acid ethyl ester

M-Clarity™ quality level = MQ300

W243213-100G-K W243213-1KG-K W243213-4KG-K
SIALW243213-100G-KEA 76.5 GBP
SIALW243213-100G-K SIALW243213-1KG-K SIALW243213-4KG-K
Ethyl decanoate, Sigma-Aldrich®
Ethyl decanoate
Formula: C₁₂H₂₄O₂
MW: 200.32 g/mol
Boiling Pt: 245
Melting Pt: –20 °C
Density: 0.862 g/cm³ (20 °C)
MDL Number: MFCD00009581
CAS Number: 110-38-3
EINECS: 203-761-9
Merck Index: 13,03811

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR