p-Cresol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ300

W233706-10KG-K W233706-1KG-K W233706-25KG-K W233706-5KG-K
SIALW233706-10KG-KEA 646 GBP
SIALW233706-10KG-K SIALW233706-1KG-K SIALW233706-25KG-K SIALW233706-5KG-K
p-Cresol, Sigma-Aldrich®
p-Cresol
Formula: C₇H₈O
Boiling Pt: 201.8 °C (1013 hPa)
Melting Pt: 32…34 °C
Density: 1.034 g/cm³ (20 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00002376
CAS Number: 106-44-5
EINECS: 203-398-6
UN: 3455
ADR: 6.1,II
Merck Index: 13,02604

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR