Butyric acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Butanoic acid

M-Clarity™ quality level = MQ100

W222119-10KG-K W222119-1KG-K W222119-25KG-K W222119-5KG-K
SIALW222119-10KG-KEA 393 GBP
SIALW222119-10KG-K SIALW222119-1KG-K SIALW222119-25KG-K SIALW222119-5KG-K
Butyric acid, Sigma-Aldrich®
Butyric acid
Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 163.5
Melting Pt: –3 °C
Density: 0.958 g/cm³ (20 °C)
Flash Pt: 67
Storage Temperature: Ambient
MDL Number: MFCD00002814
CAS Number: 107-92-6
EINECS: 203-532-3
UN: 2820
ADR: 8 III
Merck Index: 13,01593

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR