Isobutyraldehyde, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ100

W222003-1KG-K W222003-20KG-K W222003-4KG-K W222003-8KG-K
SIALW222003-1KG-KEA 124 GBP
SIALW222003-1KG-K SIALW222003-20KG-K SIALW222003-4KG-K SIALW222003-8KG-K
Isobutyraldehyde, Sigma-Aldrich®
Isobutyraldehyde
Formula: C₄H₈O
Boiling Pt: 64 °C (1013 hPa)
Melting Pt: –65 °C
Density: 0.794 g/cm³ (20 °C)
Flash Pt: –24 °C
Storage Temperature: Ambient
MDL Number: MFCD00006980
CAS Number: 78-84-2
EINECS: 201-149-6
UN: 2045
ADR: 3,II
Merck Index: 13,05171

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR