Butyl butyrate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ300

W218618-1KG-K W218618-9KG-K
SIALW218618-1KG-KEA 55 GBP
SIALW218618-1KG-K SIALW218618-9KG-K
Butyl butyrate, Sigma-Aldrich®
Butyl butyrate
Formula: C₈H₁₆O₂
Boiling Pt: 166 °C (1013 hPa)
Melting Pt: –91.5 °C
Density: 0.872 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009450
CAS Number: 109-21-7
EINECS: 203-656-8
UN: 3272
ADR: 3,III
Merck Index: 13,01555

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR