Vacuum manifolds, V20, Sigma-Aldrich®
Supplier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VM20-1SET
SIALVM20-1SETEA 235 GBP
SIALVM20-1SET
Vacuum manifolds, V20, Sigma-Aldrich®
Manifolds
The VM20 vacuum manifold is designed to provide increased efficiency and performance when used with column-based nucleic acid purification systems.
  • Dual port design allows waste to be collected in the manifold body or diverted to any auxiliary reservoir
  • Stepped design for easy access to every column
  • Endura-Luer stopcocks instead of plugs
  • Chemically resistant PP
This manifold is capable of processing 20 samples simultaneously and includes a stepped design for easy access to every column. Can be used for the isolation of circulating tumor cells byMore Product Information