Triphenylphosphine oxide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ200

T84603-100G T84603-25G
SIALT84603-100GEA 81.3 GBP
SIALT84603-100G SIALT84603-25G
Triphenylphosphine oxide, Sigma-Aldrich®
Triphenylphosphine oxide
Formula: C₁₈H₁₅OP
Boiling Pt: ∼360 °C (1013 hPa)
Melting Pt: 155…158 °C
Density: 1.2 g/cm³ (20 °C)
Flash Pt: 180 °C
MDL Number: MFCD00002080
CAS Number: 791-28-6
EINECS: 212-338-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR