3,4,5-Trimethoxyaniline, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning


MQ100

T68209-10G T68209-25G
SIALT68209-10GEA 64.9 GBP
SIALT68209-10G SIALT68209-25G
3,4,5-Trimethoxyaniline, Sigma-Aldrich®
3,4,5-Trimethoxyaniline
Formula: C₉H₁₃NO₃
MW: 183.21 g/mol
Boiling Pt: 293
Melting Pt: 110…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00008393
CAS Number: 24313-88-0
EINECS: 246-154-4
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR