2,2,2-Trifluoroethanol ≥99%, ReagentPlus®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

T63002-25G
SIALT63002-25GEA 25.2 GBP
SIALT63002-25G
2,2,2-Trifluoroethanol ≥99%, ReagentPlus®
2,2,2-Trifluoroethanol
Formula: C₂H₃F₃O
Boiling Pt: 74.1 °C (1013 hPa)
Melting Pt: –43.5 °C
Density: 1.39 g/cm³ (20 °C)
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00004672
CAS Number: 75-89-8
EINECS: 200-913-6
UN: 2929
ADR: 6.1,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR