DL-m-Tyrosine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 3-Hydroxyphenylalanine, 3-(m-Hidroxifenil)-DL-alanina, 3-(3-Hydroxyphenyl)-DL-alanine

M-Clarity™ quality level = MQ200

T3629-1G
SIALT3629-1GEA 160 GBP
SIALT3629-1G
DL-m-Tyrosine, Sigma-Aldrich®
DL-m-Tyrosine
Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 283 °C
Density: 1.456 g/cm³ (20 °C)
MDL Number: MFCD00002597
CAS Number: 775-06-4
EINECS: 212-270-9
Merck Index: 12,09971

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR