Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: N1,N1'-(Butane-1,4-diyl)bis(propane-1,3-diamine), Spermine

M-Clarity™ quality level = MQ200

S3256-1G S3256-25G S3256-5G
SIALS3256-1GEA 95.6 GBP
SIALS3256-1G SIALS3256-25G SIALS3256-5G
Spermine-(N,N'-bis(3-aminopropyl)-1,4-butanediamine), Sigma-Aldrich®
Spermine (N,N'-Bis(3-aminopropyl)-1,4-butanediamine)
Formula: C₁₀H₂₆N₄
MDL Number: MFCD00008215
CAS Number: 71-44-3
EINECS: 200-754-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR