Tin(II) 2-ethylhexanoate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

S3252-1KG S3252-100G S3252-250G
SIALS3252-1KGEA 333 GBP
SIALS3252-1KG SIALS3252-100G SIALS3252-250G
Tin(II) 2-ethylhexanoate, Sigma-Aldrich®
Tin (II) 2-ethylhexanoate
Formula: C₁₆H₃₀O₄Sn
MDL Number: MFCD00002676
CAS Number: 301-10-0
EINECS: 206-108-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR