Saccharin sodium hydrate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: o-Benzoic sulphimide sodium salt hydrate, Sodium 3-oxo-3H-benzo[d]isothiazol-2-ide 1,1-dioxide hydrate, Saccharin, sodium salt hydrate

M-Clarity™ quality level = MQ200

S1002-1KG S1002-500G
SIALS1002-1KGEA 213 GBP
SIALS1002-1KG SIALS1002-500G
Saccharin sodium hydrate, Sigma-Aldrich®
Saccharin sodium hydrate
Formula: C₇H₅NNaO₃S·xH₂O
MDL Number: MFCD00149605
CAS Number: 82385-42-0
EINECS: 204-886-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR