Strychnine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ200

S0532-100G S0532-10G S0532-25G S0532-5G
SIALS0532-100GEA 312 GBP
SIALS0532-100G SIALS0532-10G SIALS0532-25G SIALS0532-5G
Strychnine, Sigma-Aldrich®
Strychnine
Formula: C₂₁H₂₂N₂O₂
MDL Number: MFCD00005941
CAS Number: 57-24-9
EINECS: 200-319-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR