Spermidine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: N1-(3-Aminopropyl)butane-1,4-diamine, N-(3-Aminopropyl)-1,4-diaminobutane, 4-Azaoctamethylenediamine

MQ200

S0266-1G S0266-25G S0266-5G
SIALS0266-1GEA 80.9 GBP
SIALS0266-1G SIALS0266-25G SIALS0266-5G
Spermidine, Sigma-Aldrich®
Spermidine
Formula: C₇H₁₉N₃
MW: 145.25 g/mol
Boiling Pt: 246
Melting Pt: 20…25 °C
Density: 0.925 g/cm³ (20 °C)
MDL Number: MFCD00008229
CAS Number: 124-20-9
EINECS: 204-689-0
Merck Index: 12,08893

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR