Polyvinylpyrrolidone, M.W. ∼ 40,000 (K30), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ200

PVP40-100G PVP40-1KG PVP40-500G PVP40-50G PVP40-5KG
SIALPVP40-100GEA 74.2 GBP
SIALPVP40-100G SIALPVP40-1KG SIALPVP40-500G SIALPVP40-50G SIALPVP40-5KG
Polyvinylpyrrolidone, M.W. ∼ 40,000 (K30), Sigma-Aldrich®
Polyvinylpyrrolidone
Formula: (C₆H₉NO)n
Storage Temperature: Ambient
MDL Number: MFCD01076626
CAS Number: 9003-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR