5-Methylphenazinium methyl sulphate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: PMS, 5-Methylphenazin-5-ium methyl sulphate, Phenazine methosulphate, Phenazine methylsulphate

M-Clarity™ quality level = MQ200

P9625-500MG P9625-5G P9625-10G P9625-1G P9625-25G
SIALP9625-500MGEA 43 GBP
SIALP9625-500MG SIALP9625-5G SIALP9625-10G SIALP9625-1G SIALP9625-25G
5-Methylphenazinium methyl sulphate, Sigma-Aldrich®
5-Methylphenazinium methyl sulphate
Formula: C₁₄H₁₄N₂O₄S
Melting Pt: 198 °C
Storage Temperature: Freezer
MDL Number: MFCD00011923
CAS Number: 299-11-6
EINECS: 206-072-1
Merck Index: 12,06185

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR