Prednisolone, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Prednisolonum

M-Clarity™ quality level = MQ200

P6004-1G P6004-100MG P6004-10G P6004-25G P6004-5G
SIALP6004-1GEA 66.2 GBP
SIALP6004-1G SIALP6004-100MG SIALP6004-10G SIALP6004-25G SIALP6004-5G
Prednisolone, Sigma-Aldrich®
Prednisolone
Formula: C₂₁H₂₈O₅
Boiling Pt: 571 °C (1013 hPa)
Melting Pt: 246 °C
Density: 1.212 g/cm³ (20 °C)
MDL Number: MFCD00003649
CAS Number: 50-24-8
EINECS: 200-021-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR