Palmitic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ200

P5585-100G P5585-10G P5585-25G
SIALP5585-100GEA 393 GBP
SIALP5585-100G SIALP5585-10G SIALP5585-25G
Palmitic acid, Sigma-Aldrich®
Palmitic acid
Formula: C₁₆H₃₂O₂
Boiling Pt: 352 °C (1013 hPa)
Melting Pt: 61…63 °C
Density: 0.853 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002747
CAS Number: 57-10-3
EINECS: 200-312-9
Merck Index: 13,07064

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR