N-Octyl-β-D-glucopyranoside, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Octylglucopyranoside, OG, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(octyloxy)tetrahydro-2H-pyran-3,4,5-triol, n-Octyl β-D-glucopyranoside, 1-O-n-Octyl-β-D-glucopyranoside, Octyl-β-D-glucopyranoside

MQ300

O8001-100G O8001-100MG O8001-10G O8001-1G O8001-250MG O8001-25G O8001-500MG O8001-5G
SIALO8001-100GEA 10750 GBP
SIALO8001-100G SIALO8001-100MG SIALO8001-10G SIALO8001-1G SIALO8001-250MG SIALO8001-25G SIALO8001-500MG SIALO8001-5G
N-Octyl-β-D-glucopyranoside, Sigma-Aldrich®
N-Octyl-β-D-glucopyranoside
Formula: C₁₄H₂₈O₆
MW: 292.37 g/mol
Melting Pt: 110
Storage Temperature: Freezer
MDL Number: MFCD00063288
CAS Number: 29836-26-8
EINECS: 249-887-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR