n-Octadecane, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Octadecane

O652-100G O652-25G O652-500G
SIALO652-100GEA 110 GBP
SIALO652-100G SIALO652-25G SIALO652-500G
n-Octadecane, Sigma-Aldrich®
n-Octadecane
Formula: H₃C(CH₂)₁₆CH₃
Boiling Pt: 316.1 °C (1013 hPa)
Melting Pt: 26…29 °C
Density: 0.7765 g/cm³ (20 °C)
Flash Pt: 165 °C
Storage Temperature: Ambient
MDL Number: MFCD00009007
CAS Number: 593-45-3
EINECS: 209-790-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR