L(+)-Ornithine hydrochloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ200

O4386-100G O4386-25G
SIALO4386-100GEA 403 GBP
SIALO4386-100G SIALO4386-25G
L(+)-Ornithine hydrochloride, Sigma-Aldrich®
L(+)-Ornithine hydrochloride
Formula: C₅H₁₂N₂O₂·HCl
MDL Number: MFCD00064562
CAS Number: 3184-13-2
EINECS: 221-678-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR