1-Naphthylacetic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Planofix, 1-Naphthaleneacetic acid

M-Clarity™ quality level = MQ200

N9272-500G
SIALN9272-500GEA 146 GBP
SIALN9272-500G
1-Naphthylacetic acid, Sigma-Aldrich®
1-Naphthylacetic acid
Formula: C₁₂H₁₀O₂
Boiling Pt: 352 °C (1013 hPa)
Melting Pt: 129…131 °C
Density: ∼1.263 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004046
CAS Number: 86-87-3
EINECS: 201-705-8
Merck Index: 12,06458

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR