3-Methyl-1H-pyrazole, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 3-Methylpyrazole

M-Clarity™ quality level = MQ100

M75802-25G M75802-5G
SIALM75802-25GEA 61.9 GBP
SIALM75802-25G SIALM75802-5G
3-Methyl-1H-pyrazole, Sigma-Aldrich®
3-Methyl-1H-pyrazole
Formula: C₄H₆N₂
MDL Number: MFCD00005240
CAS Number: 1453-58-3
EINECS: 215-925-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR