Mycophenolic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning


M-Clarity™ quality level = MQ300

M5255-250MG M5255-50MG
SIALM5255-250MGEA 408 GBP
SIALM5255-250MG SIALM5255-50MG
Mycophenolic acid, Sigma-Aldrich®
Mycophenolic acid
Formula: C₁₇H₂₀O₆
Storage Temperature: Refrigerator
MDL Number: MFCD00036814
CAS Number: 24280-93-1
EINECS: 246-119-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR