2-Mercaptobenzothiazole, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 2-Benzothiazolethiol, Mercapto-2 benzothiazole, Benzo[d]thiazole-2(3H)-thione, Benzothiazole-2-thiol, MBT

M-Clarity™ quality level = MQ200

M3302-100G M3302-1KG M3302-5G
SIALM3302-100GEA 64 GBP
SIALM3302-100G SIALM3302-1KG SIALM3302-5G
2-Mercaptobenzothiazole, Sigma-Aldrich®
2-Mercaptobenzothiazole
Formula: C₇H₅NS₂
Boiling Pt: 302 °C (1013 hPa)
Melting Pt: 177…182 °C
Density: 1.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005781
CAS Number: 149-30-4
EINECS: 205-736-8
UN: 3077
ADR: 9,III
Merck Index: 13,05893

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR