γ-(N-Morpholino)propanesulphonic acid (MOPS), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid

MQ300

M3183-100G M3183-1KG M3183-25G M3183-500G M3183-6X100G M3183-6X500G
SIALM3183-100GEA 165 GBP
SIALM3183-100G SIALM3183-1KG SIALM3183-25G SIALM3183-500G SIALM3183-6X100G SIALM3183-6X500G
γ-(N-Morpholino)propanesulphonic acid (MOPS), Sigma-Aldrich®
γ-(N-Morpholino)propanesulphonic acid (MOPS)
Formula: C₇H₁₅NO₄S
MW: 209.27 g/mol
Melting Pt: > 250 °C
Flash Pt: 110 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00006183
CAS Number: 1132-61-2
EINECS: 214-478-5
Merck Index: 13,06288

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR