Magnesium sulphate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: MgSO4, Magnesium sulphate anhydrous

M-Clarity™ quality level = MQ200

M2643-1KG M2643-500G
SIALM2643-1KGEA 306 GBP
SIALM2643-1KG SIALM2643-500G
Magnesium sulphate, Sigma-Aldrich®
Magnesium sulphate
Formula: MgSO₄
Melting Pt: 1127 °C
Density: 2.65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011110
CAS Number: 7487-88-9
EINECS: 231-298-2
Merck Index: 12,05731
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR