Isonicotinic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning


M-Clarity™ quality level = MQ200

I17508-100G I17508-500G I17508-5G
SIALI17508-100GEA 66.5 GBP
SIALI17508-100G SIALI17508-500G SIALI17508-5G
Isonicotinic acid, Sigma-Aldrich®
Isonicotinic acid
Formula: C₆H₅NO₂
Boiling Pt: 396 °C (1013 hPa)
Melting Pt: 315…319 °C
Storage Temperature: Ambient
MDL Number: MFCD00006429
CAS Number: 55-22-1
EINECS: 200-228-2
Merck Index: 13,05206

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR