Iodoacetamide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning


M-Clarity™ quality level = MQ300

I1149-100G I1149-25G I1149-5G
SIALI1149-100GEA 1110 GBP
SIALI1149-100G SIALI1149-25G SIALI1149-5G
Iodoacetamide, Sigma-Aldrich®
Iodoacetamide
Formula: ICH₂CONH₂
Boiling Pt: 266 °C (1013 hPa)
Melting Pt: 91…94 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008028
CAS Number: 144-48-9
EINECS: 205-630-1
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR