2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Iodonitrotetrazolium chloride, p-Iodonitrotetrazolium violet, INT, 3-(4-Iodophenyl)-2-(4-nitrophenyl)-5-phenyl-2H-tetrazolium chloride, 2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyl-2H-tetrazolium chloride, 2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyl-2H-tetrazol-3-ium chloride, Iodonitrotetrazolium violet, 3-(4-Iodophenyl)-2-(4-nitrophenyl)-5-phenyltetrazolium chloride

M-Clarity™ quality level = MQ200

I10406-1G I10406-25G I10406-5G
SIALI10406-1GEA 101 GBP
SIALI10406-1G SIALI10406-25G SIALI10406-5G
2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride, Sigma-Aldrich®
2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride
Formula: C₁₉H₁₃ClIN₅O₂
Melting Pt: 227…229 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011961
CAS Number: 146-68-9
EINECS: 205-676-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR