Hydroxypropyl methylcellulose (Hypromellose), 4,000 mPa.s (2% in H2O), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Methocel K100, Hydroxypropyl methyl cellulose

M-Clarity™ quality level = MQ200

H3785-100G H3785-250G H3785-25G
SIALH3785-100GEA 378 GBP
SIALH3785-100G SIALH3785-250G SIALH3785-25G
Hydroxypropyl methylcellulose (Hypromellose), 4,000 mPa.s (2% in H2O), Sigma-Aldrich®
Hydroxypropyl methyl cellulose (Hypromellose)
Formula: C₃H₈O₂·xCH₄O
MDL Number: MFCD00131360
CAS Number: 9004-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR