Riboflavine-5'-phosphate sodium salt dihydrate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

F1392-25MG
SIALF1392-25MGEA 155 GBP
SIALF1392-25MG
Riboflavine-5'-phosphate sodium salt dihydrate, Sigma-Aldrich®
Riboflavine-5'-phosphate sodium salt dihydrate
Formula: C₁₇H₂₀N₄NaO₉P·2H₂O
Storage Temperature: Freezer
MDL Number: MFCD00150993
CAS Number: 6184-17-4
EINECS: 204-988-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR