m-Ethyltoluene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ100

E49606-10G E49606-25G
SIALE49606-10GEA 253 GBP
SIALE49606-10G SIALE49606-25G
m-Ethyltoluene, Sigma-Aldrich®
m-Ethyltoluene
Formula: C₉H₁₂
Boiling Pt: 158…160 °C (1013 hPa)
Melting Pt: –95.5 °C
Density: 0.869 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009259
CAS Number: 620-14-4
EINECS: 210-626-8
UN: 3295
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR