Tricaine methanesulphonate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ100

E10521-10G E10521-50G
SIALE10521-10GEA 63.6 GBP
SIALE10521-10G SIALE10521-50G
Tricaine methanesulphonate, Sigma-Aldrich®
Tricaine methanesulphonate
Formula: C₁₀H₁₅NO₅S
MDL Number: MFCD00013176
CAS Number: 886-86-2
EINECS: 212-956-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR