Ethanolamine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ300

E0135-100ML E0135-500ML
SIALE0135-100MLEA 65 GBP
SIALE0135-100ML SIALE0135-500ML
Ethanolamine, Sigma-Aldrich®
Ethanolamine
Formula: NH₂CH₂CH₂OH
Boiling Pt: 171 °C (1013 hPa)
Melting Pt: 10.5 °C
Density: 1.0164 g/cm³ (20 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00008183
CAS Number: 141-43-5
EINECS: 205-483-3
UN: 2491
ADR: 8,III
Merck Index: 13,03762
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82bfb6ea7cf5dde341c60ff9cf752bc5c422eb28&vid=b387e8c39029da4ea050ba6dcfb6123d81ab03c8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR