3,4-Dichlorophenol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ200

D70406-25G D70406-5G
SIALD70406-25GEA 119 GBP
SIALD70406-25G SIALD70406-5G
3,4-Dichlorophenol, Sigma-Aldrich®
3,4-Dichlorophenol
Formula: C₆H₄Cl₂O
Boiling Pt: 247 °C (1013 hPa)
Melting Pt: 66…68 °C
Density: 1.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002258
CAS Number: 95-77-2
EINECS: 202-450-5
UN: 2020
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR