2,4-Dichlorobenzylamine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: (2,4-Dichlorophenyl)methanamine

D58406-5G
SIALD58406-5GEA 39.2 GBP
SIALD58406-5G
2,4-Dichlorobenzylamine, Sigma-Aldrich®
2,4-Dichlorobenzylamine
Formula: C₇H₇Cl₂N
MW: 176.04 g/mol
Boiling Pt: 83
Density: 1.33 g/cm³ (20 °C)
MDL Number: MFCD00008109
CAS Number: 95-00-1
EINECS: 202-382-6
UN: 2735
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR