ω,4-Dibromoacetophenone, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ200

D38308-100G D38308-10G D38308-50G
SIALD38308-100GEA 175 GBP
SIALD38308-100G SIALD38308-10G SIALD38308-50G
ω,4-Dibromoacetophenone, Sigma-Aldrich®
ω,4-Dibromoacetophenone
Formula: C₈H₆Br₂O
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 107…108 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000200
CAS Number: 99-73-0
EINECS: 202-783-6
UN: 3261
ADR: 8,II
Merck Index: 13,01412

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR