2,5-Diphenyloxazole, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: PPO, 2,5-Diphenyl-1,3-oxazole

M-Clarity™ quality level = MQ200

D210404-100G D210404-25G D210404-500G
SIALD210404-100GEA 202 GBP
SIALD210404-100G SIALD210404-25G SIALD210404-500G
2,5-Diphenyloxazole, Sigma-Aldrich®
2,5-Diphenyloxazole
Formula: C₁₅H₁₁NO
Boiling Pt: 360 °C (1013 hPa)
Melting Pt: 70…72 °C
Density: 1.094 g/cm³ (99 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005306
CAS Number: 92-71-7
EINECS: 202-181-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR