Calcium carbonate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Marble

M-Clarity™ quality level = MQ200

C5929-100G C5929-500G
SIALC5929-100GEA 41.6 GBP
SIALC5929-100G SIALC5929-500G
Calcium carbonate, Sigma-Aldrich®
Calcium carbonate
Formula: CaCO₃
Melting Pt: 825 °C
Density: 2.71 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010906
CAS Number: 471-34-1
EINECS: 207-439-9
Merck Index: 13,01658

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR