4-Chloro-m-cresol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 4-Chloro-3-methylphenol, 2-Chloro-5-hydroxytoluene, 1-Chloro-4-hydroxy-2-methylbenzene, p-Chloro m-cresol

M-Clarity™ quality level = MQ100

C55402-5G C55402-100G C55402-500G
SIALC55402-5GEA 46.2 GBP
SIALC55402-5G SIALC55402-100G SIALC55402-500G
4-Chloro-m-cresol, Sigma-Aldrich®
4-Chloro-m-cresol
Formula: C₇H₇ClO
MW: 142.58 g/mol
Boiling Pt: 235
Melting Pt: 63…65 °C
Density: 0.9 g/cm³ (20 °C)
Flash Pt: 118
Storage Temperature: Ambient
MDL Number: MFCD00002323
CAS Number: 59-50-7
EINECS: 200-431-6
UN: 3437
ADR: 6.1 II
Merck Index: 13,02152

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR