(1R,3S)-(+)-Camphoric acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: D(+)-Camphoric acid, (1R,3S)-1,2,2-Trimethylcyclopentane-1,3-dicarboxylic acid, Levocarnitine Impurity B

M-Clarity™ quality level = MQ200

C409-100G C409-5G
SIALC409-100GEA 91.9 GBP
SIALC409-100G SIALC409-5G
(1R,3S)-(+)-Camphoric acid, Sigma-Aldrich®
(1R,3S)-(+)-Camphoric acid
Formula: C₁₀H₁₆O₄
MDL Number: MFCD00001375
CAS Number: 124-83-4
EINECS: 204-715-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR