(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propanesulphonate), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: (3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxypropanesulphonate), CHAPSO

MQ200

C3649-1G C3649-25G C3649-500MG C3649-5G
SIALC3649-1GEA 198 GBP
SIALC3649-1G SIALC3649-25G SIALC3649-500MG SIALC3649-5G
(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propanesulphonate), Sigma-Aldrich®
(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propanesulphonate)
Formula: C₃₂H₅₈N₂O₈S
MW: 630.89 g/mol
Melting Pt: 184…186 °C
Storage Temperature: Refrigerator
CAS Number: 82473-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR