5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning


M-Clarity™ quality level = MQ200

B6777-100MG B6777-500MG
SIALB6777-100MGEA 155 GBP
SIALB6777-100MG SIALB6777-500MG
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate), Sigma-Aldrich®
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate)
Formula: C₁₅H₁₅BrClN₂O₄P
Storage Temperature: Freezer
MDL Number: MFCD00040636
CAS Number: 6578-06-9
EINECS: 229-506-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR