ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate]), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: ABTS, Ammonium 2,2'-(hydrazine-1,2-diylidene)bis(3-ethyl-2,3-dihydrobenzo[d]thiazole-6-sulphonate), 2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulphonic acid) diammonium salt

M-Clarity™ quality level = MQ200

A9941-100TAB A9941-50TAB A9941-5TAB
SIALA9941-100TABEA 469 GBP
SIALA9941-100TAB SIALA9941-50TAB SIALA9941-5TAB
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate]), Sigma-Aldrich®
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate])
Formula: C₁₈H₂₄N₆O₆S₄
Storage Temperature: Refrigerator
MDL Number: MFCD00010404
CAS Number: 30931-67-0
EINECS: 250-396-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR