2-(Aminomethyl)pyridine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 2-Pyridylmethylamine, 2-Picolylamine, Pyridin-2-ylmethanamine, (2-Pyridyl)methylamine, 2-Picolamine

M-Clarity™ quality level = MQ100

A65204-25G A65204-100G A65204-5G
SIALA65204-25GEA 79 GBP
SIALA65204-25G SIALA65204-100G SIALA65204-5G
2-(Aminomethyl)pyridine, Sigma-Aldrich®
2-(Aminomethyl)pyridine
Formula: C₆H₈N₂
Boiling Pt: 197 °C (1013 hPa)
Melting Pt: 81 °C
Density: 1.0583 g/cm³ (20 °C)
MDL Number: MFCD00006360
CAS Number: 3731-51-9
EINECS: 223-090-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR