1-(2-Aminoethyl)piperazine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ100

A55209-100G A55209-500G
SIALA55209-100GEA 37.8 GBP
SIALA55209-100G SIALA55209-500G
1-(2-Aminoethyl)piperazine, Sigma-Aldrich®
1-(2-Aminoethyl)piperazine
Formula: C₆H₁₅N₃
MDL Number: MFCD00005971
CAS Number: 140-31-8
EINECS: 205-411-0
UN: 2815
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR