O-Acetylsalicylic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning


M-Clarity™ quality level = MQ200

A3160-1VL
SIALA3160-1VLEA 213 GBP
SIALA3160-1VL
O-Acetylsalicylic acid, Sigma-Aldrich®
o-Acetylsalicylic acid
Formula: 2-(CH₃CO₂)C₆H₄CO₂H
Boiling Pt: 140 °C (1013 hPa)
Melting Pt: 138…140 °C
Density: 1.39 g/cm³ (20 °C)
Flash Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD00002430
CAS Number: 50-78-2
EINECS: 200-064-1
Merck Index: 13,00856
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fcb60a4ee505b596cef38216a85e034985091e0&vid=5f8d5cfeb3039783869a9320ec840b8ec12c33ea&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR