Agar, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Agar Agar

MQ200

A1296-100G A1296-10KG A1296-1KG A1296-25KG A1296-500G A1296-5KG
SIALA1296-100GEA 98.5 GBP
SIALA1296-100G SIALA1296-10KG SIALA1296-1KG SIALA1296-25KG SIALA1296-500G SIALA1296-5KG
Agar, Sigma-Aldrich®
Agar
Formula: C₁₂nH₁₈nOn₉n
Melting Pt: 90
Storage Temperature: Ambient
MDL Number: MFCD00081288
CAS Number: 9002-18-0
EINECS: 232-658-1
Merck Index: 13,00184
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36877cbcfaca6a0ace648ee20ae51b3467f2748c&vid=4295a0dcbc54bbbd374a5fb934297574c6848d32&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR